missing translation for 'onlineSavingsMsg'
Learn More

Cesium chloride, ultra dry, 99.998% (metals basis)

Código de producto. 11383867
Change view
Click to view available options
Quantity:
5 g
25 g
Tamaño de la unidad:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity unitSize
11383867 25 g 25g
11373867 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11383867 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor 035690.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 7647-17-8 | ClCs | 168.36 g/mol

It is used as a precursor for cesium metal, it act as a phase transfer catalyst, for molecular genetic applications used in preparing nucleic acids for subsequent hybridization or cloning experiments. It a useful compound for density gradient centrifugation, for fractionation of ribosomal subunits, proteins, glycoproteins, and viruses, evacuation of radio tubes, in the preparation of electrically conducting glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name cesium;chloride
SMILES [Cl-].[Cs+]

Especificaciones

Boiling Point 1303°C
Melting Point 646°C
Physical Form Beads
Quantity 25 g
Merck Index 14,2011
Solubility Information Soluble in water.
Formula Weight 168.36
Percent Purity 99.998%
Odor Odorless
Packaging Ampule
Refractive Index 1.6418
Sensitivity Hygroscopic
Density 3.99 g/mL
Assay (metals basis)
Chemical Name or Material Cesium chloride, Ultra dry
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.