missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Chloride, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
Description
- Cesium chloride is typically used for density gradient work and for the purification of virus/phage, nucleic acids and nucleoproteins.
- It is used for the preparation of electrically conducting glasses, used to make solutions for the separation of RNA from DNA by density gradient centrifugation.
- Cesium chloride (CsCl) is commonly used in the separation of DNA and RNA.
- Cesium chloride , an inorganic compound, is an important source of cesium ions in a variety of applications.
Specifications
Specifications
| Boiling Point | 1303°C (literature) |
| Melting Point | 646°C (literature) |
| Color | White |
| pH | 5.5 to 6.5 (1% aq soln) |
| Physical Form | Powder |
| Quantity | 100 g |
| Solubility Information | Freely soluble in water (500mg/mL),(50% aqueous-clear, colorless solution); soluble in ethanol, alcohol |
| Grade | 0.99999 |
| Refractive Index | 1.642(Lit.) |
| Density | 3.99g/mL(Lit.) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction