missing translation for 'onlineSavingsMsg'
Learn More

Cesium chloride, 99.9% (metals basis)

Product Code. 11336717
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336717 100 g 100g
11326717 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336717 Supplier Thermo Scientific Alfa Aesar Supplier No. 010018.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-17-8 | ClCs | 168.36 g/mol

Analytical chemistry reagent for detecting inorganic ions. Source of cesium metal. Phase transfer catalyst in organic chemistry. Used for density gradient work to separate and purify proteins (isopycnic centrifugation) including chromosomal, plasmid and organelle (mitochondrial or plasmid) DNAs, and different RNA types. Preparation of electrically conducting glasses and screens of cathode ray tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1303°C
Melting Point 646°C
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in water and concentrated hydrochloric acid.
Formula Weight 168.36
Refractive Index 1.6418
Sensitivity Hygroscopic
Density 3.99
Chemical Name or Material Cesium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.