missing translation for 'onlineSavingsMsg'
Learn More

Cesium carbonate, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals

Catalog Number 11376478
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
534-17-8
CCs2O3
325.82
MFCD00010957
FJDQFPXHSGXQBY-UHFFFAOYSA-L
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
10796
[Cs+].[Cs+].[O-]C([O-])=O
This item is not returnable. View return policy
534-17-8
CCs2O3
325.82
MFCD00010957
FJDQFPXHSGXQBY-UHFFFAOYSA-L
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
10796
[Cs+].[Cs+].[O-]C([O-])=O

Cesium carbonate is the precursor for other cesium compounds. It acts as a base in sensitive organic reactions. Suzuki, Heck and Sonogashira synthesis are some of the important reactions involving cesium carbonate. It finds use in solar cells as it increases the power conversion efficiency of cells through the transfer of electrons. It is also used in the production of special optical glasses, petroleum catalytic additives, special ceramics, and in sulfuric acid industry. It is useful in the N-alkylation (of sulfonamides, β-lactams, indoles, heterocycles, and several sensitive nitrogen compounds), carbamination of amines, carbonylation of alcohols, and aerobic oxidation of alcohols into carbonyl compounds without over oxidations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 610°C (decomposition)
Color White
Physical Form Powder/Granules
Quantity 5 g
Assay Percent Range 99.994% (Metal basis)
Beilstein 4546405
Solubility Information Highly soluble in water. Soluble in alcohol,dimethyl sulfoxide and N,N-dimethylformamide. Insoluble in toluene and p-xylene.
IUPAC Name dicesium;carbonate
Formula Weight 325.82
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder
Density 4.072
Chemical Name or Material Cesium carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'