missing translation for 'onlineSavingsMsg'
Learn More

Cesium bromide, 99.999% (metals basis)

Product Code. 11315009
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11315009 10 g 10g
11305009 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11315009 Supplier Thermo Scientific Alfa Aesar Supplier No. 087639.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7787-69-1 | BrCs | 212.81 g/mol

Cesium bromide is a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-69-1
Molecular Formula BrCs
Molecular Weight (g/mol) 212.81
MDL Number MFCD00010954
InChI Key LYQFWZFBNBDLEO-UHFFFAOYSA-M
Synonym cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide
PubChem CID 24592
IUPAC Name cesium;bromide
SMILES [Br-].[Cs+]

Specifications

Boiling Point 1300°C
Melting Point 636°C
Color White
Quantity 10 g
Linear Formula CsBr
Merck Index 14,2009
Solubility Information Soluble in water. (55g/100 mL (20°C))
Formula Weight 212.81
Percent Purity 99.999%
Odor Odorless
Refractive Index 1.6984
Sensitivity Hygroscopic
Density 4.44 g/mL
Assay (metals basis)
Chemical Name or Material Cesium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.