missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Bicarbonate, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Cesium Bicarbonate |
| CAS | 29703-01-3 |
| Molecular Formula | C H O3 . Cs |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/CH2O3.Cs/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1 |
| SMILES | [Cs+].OC(=O)[O-] |
| IUPAC Name | cesium;hydrogen carbonate |
| Molecular Weight (g/mol) | 193.922 |
| Formula Weight | 193.898 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction