missing translation for 'onlineSavingsMsg'
Learn More

Cesium Acetate, Technical grade, ≥95%, Honeywell™ Fluka™

Product Code. 15691720 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Poly Bottle
Unit Size:
250g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15691720 25 g Poly Bottle 25g
15681720 250 g Poly Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15691720 Supplier Honeywell-Fluka Supplier No. 20079425G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 3396-11-0
Molecular Formula C2H3CsO2
Molecular Weight (g/mol) 191.949
MDL Number MFCD00013056
InChI Key ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
Synonym cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion
PubChem CID 5152919
IUPAC Name cesium;acetate
SMILES CC(=O)[O-].[Cs+]

Specifications

Melting Point 194°C
Color White
Physical Form Solid
Quantity 25 g
Linear Formula CH3COOCs
UN Number NONH for all modes of transport
Beilstein 3595637
Formula Weight 191.95g/mol
Percent Purity ≥95%
Grade Technical
Packaging Poly Bottle
Chemical Name or Material Cesium Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.