missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CES7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19520-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody can recognize the isoform1 and isoform2.
CES7 is involved in the detoxification of xenobiotics and in the activation of ester and amide prodrugs.Specifications
| CES7 | |
| Polyclonal | |
| Unconjugated | |
| CES5A | |
| Carboxylesterase 7, CAUXIN, CES4C1, CES5, CES7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 221223 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.46 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6NT32 | |
| CES5A | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction