missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CES7 Polyclonal Antibody

CES7 Polyclonal Antibody
Brand: Thermo Scientific PA561561
This item is not returnable.
View return policy
Description
CES7 Polyclonal Antibody for Western Blot, IHC
Specifications
| CES7 | |
| Polyclonal | |
| Unconjugated | |
| CES5A | |
| 1700081L16Rik; 1700122C07Rik; BB081581; carboxylesterase 5; carboxylesterase 5A; carboxylesterase 7; carboxylesterase-like urinary excreted protein homolog; CAUXIN; CES4C1; CES5; Ces5a; CES7; epididymis luminal protein 126; Gm503; HEL126 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 221223 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NT32 | |
| CES5A | |
| Recombinant protein corresponding to Human CES7. Recombinant protein control fragment (Product #RP-98685). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction