missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CES7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526164
This item is not returnable.
View return policy
Description
CES7 Polyclonal Antibody for Western Blot, IHC (P)
CES7 is involved in the detoxification of xenobiotics and in the activation of ester and amide prodrugs.
Specifications
| CES7 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6NT32 | |
| CES5A | |
| KLH conjugated synthetic peptide between 244-270 amino acids from the central region of human CES7. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| CES5A | |
| 1700081L16Rik; 1700122C07Rik; BB081581; carboxylesterase 5; carboxylesterase 5A; carboxylesterase 7; carboxylesterase-like urinary excreted protein homolog; CAUXIN; CES4C1; CES5; Ces5a; CES7; epididymis luminal protein 126; Gm503; HEL126 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 221223 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction