missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CES4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524203
This item is not returnable.
View return policy
Description
CES4 Polyclonal Antibody for Western Blot, IHC (P)
Has no esterase activity (Probable).
Specifications
| CES4 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9UKY3 | |
| CES1P1 | |
| KLH conjugated synthetic peptide between 47-74 amino acids from the N-terminal region of human CES4. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| CES1P1 | |
| carboxylesterase 1 pseudogene 1; CES1A2; CES1A3; CES1P1; CES4; CESR; PCE-3 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 51716 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction