missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CES3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14587-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody recognizes both isoforms with MW 60-62 kDa and 24 kDa.
CES3 major lipase in white adipose tissue. Involved in the metabolism of xenobiotics and of natural substrates. Hydrolyzes triacylglycerols and monoacylglycerols, with a preference for monoacylglycerols. The susceptibility of the substrate increases with decreasing acyl chain length of the fatty acid moiety. Catalyzes the synthesis of fatty acid ethyl esters.Specifications
| CES3 | |
| Polyclonal | |
| Unconjugated | |
| CES3 | |
| carboxylesterase 3, CES3, ES31 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 104158, 23491 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6UWW8, Q8VCT4 | |
| Ces1d, CES3 | |
| CES3 Fusion Protein Ag6113 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction