Learn More
Ceruloplasmin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21131-1-AP-150UL
 View more versions of this product
Product Code. 16860735
  • £349.00 / 150µL
Description
The protein encoded by this gene is a metalloprotein that binds most of the copper in plasma and is involved in the peroxidation of Fe (II)transferrin to Fe (III) transferrin. Mutations in this gene cause aceruloplasminemia, which results in iron accumulation and tissue damage, and is associated with diabetes and neurologic abnormalities.
Specifications
Specifications
Ceruloplasmin
Polyclonal
Unconjugated
CP
Ceruloplasmin, ceruloplasmin (ferroxidase), CP, CP 2, Ferroxidase
Rabbit
Antigen Affinity Chromatography
RUO
12870, 1356
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P00450, Q61147
CP
Ceruloplasmin Fusion Protein Ag15535
150 μL
Primary
Human, Mouse