missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ceruloplasmin Monoclonal Antibody (C1)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543891
This item is not returnable.
View return policy
Description
Ceruloplasmin Monoclonal Antibody for Western Blot, IHC (P)
Ceruloplasmin encodes a protein that is a metalloprotein that binds most of the copper in plasma and is involved in the peroxidation of Fe(II)transferrin to Fe(III) transferrin. Mutations in this gene cause aceruloplasminemia, which results in iron accumulation and tissue damage, and is associated with diabetes and neurologic abnormalities.
Specifications
| Ceruloplasmin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P00450 | |
| CP | |
| Recombinant protein Ceruloplasmin. The antigen corresponds to amino acid range 789-1065 of the target protein. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| C1 | |
| Unconjugated | |
| CP | |
| bilitranslocase; CERP; Ceruloplasmin; ceruloplasmin (ferroxidase); CP; CP-2; D3Ertd555e; ferroxidase | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1356 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction