missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Cerulenin

Product Code. 16430748
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16430748 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16430748 Supplier MedChemExpress Supplier No. HYA0210
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cerulenin, a potent, natural inhibitor of fatty acid synthase (FASN), is an epoxide produced by the fungus Cephalosporium caeruleus. Cerulenin inhibits topoisomerase I catalytic activity and augments SN-38-induced apoptosis. Cerulenin has antifungal and antitumor activies.

Specifications

Content And Storage -20°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Molecular Weight (g/mol) 223.27 g/mol
Shipping Condition Room temperature
For Use With (Application) Metabolism-sugar/lipid metabolism
CAS 17397-89-6
Product Type Natural Products
Purity 98.0%
Solubility DMSO : 100 mg/mL (447.89 mM∣Need ultrasonic)
Target Fatty Acid Synthase (FASN)∣Fungal∣Antibiotic
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.