missing translation for 'onlineSavingsMsg'
Learn More

Paragon Scientific™ Certified Viscosity Check Oil, 5W30

The viscosity values for the certified viscosity check oils have been tested and certified at typical blend targets. These check oils are for the validating the performance of Cold-Cranking Simulator and Kinematic Viscometers by measuring commercially available formulated engine oils

Brand:  Paragon Scientific™ CVCO5W30

Additional Details : Weight : 0.90000kg

 View more versions of this product

Product Code. 16080192

  • £45.20 / 500mL
Estimated Shipment: 19-12-2023
to see stock.
Add to basket

Description

Description

  • Certified in accordance to ISO 17025 / ISO 17034
  • Kinematic Viscosity mm2/s (cSt) in accordance with ASTM D445
  • Density g/mL in accordance with ASTM D4052
  • CCS Dynamic Viscosity mPa.s (cP) in accordance with ASTM D5293
  • Viscosity Index in accordance with ASTM D2270
  • Standard Pack Size 500mL and 5L available
  • Fully traceable to National Standards
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
Specifications

Specifications

ISO 17025 / ISO 17034
500 mL
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Nominal values quoted. Certified Values will vary from batch to batch.