Learn More
Paragon Scientific™ Certified Viscosity Check Oil, 15W40
The viscosity values for the certified viscosity check oils have been tested and certified at typical blend targets. These check oils are for the validating the performance of Cold-Cranking Simulator and Kinematic Viscometers by measuring commercially available formulated engine oils
Brand: Paragon Scientific™ CVCO15W40-5L
Additional Details : Weight : 5.00000kg
Description
- Certified in accordance to ISO 17025 / ISO 17034
- Kinematic Viscosity mm2/s (cSt) in accordance with ASTM D445
- Density g/mL in accordance with ASTM D4052
- CCS Dynamic Viscosity mPa.s (cP) in accordance with ASTM D5293
- Viscosity Index in accordance with ASTM D2270
- Standard Pack Size 500mL and 5L available
- Fully traceable to National Standards
- Tamper evident security packaging
- 2 Year shelf life
- Manufactured in the United Kingdom
Specifications
ISO 17025 / ISO 17034 | |
5 L |
Nominal values quoted. Certified Values will vary from batch to batch.