missing translation for 'onlineSavingsMsg'
Learn More

Certified pH Buffer Solution, UKAS ISO 17025 / ISO 17034 Certified, 25 °C, Paragon Scientific™

Product Code. 16060280
Click to view available options
pH:
4
7
10
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 16060280

Brand: Paragon Scientific PH425CF

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifically used for verification and calibration checks of pH meter instruments. Color-coded to allow for easy identification of the pH required, and include anti-fungal preservatives / mould inhibitors. pH values are traceable to National Institute Standards and Technology (NIST.)

  • Certified solutions for 25 °C and are accurate within ± 0.01
  • Certified pH buffer solutions are fully traceable to NIST and international protocols
  • Sold as colour-coded solutions for easy-identification purposes
  • Sold with anti-fungal preservatives / mould inhibitors
  • Supplied with certificate, Safety Data Sheet (SDS) and correlation table between pH and temperature
  • Supplied in 500 mL, tamper-evident containers. For other volumes, please contact us direct.
  • 12 month shelf life per solution

Specifications

Color Red
Certificate of Analysis UKAS ISO 17025 / ISO 17034
pH 4

Nominal values quoted. Certified Values will vary from batch to batch.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.