missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CERT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143369
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. Positive control: A431, HeLa.
CERT (GPBP) is a GC-rich promotor-specific transactivating transcription factor. This protein is suggested to play a role in the development of atherosclerosis.
Specifications
| CERT | |
| Polyclonal | |
| Unconjugated | |
| CERT1 | |
| 2810404O15Rik; 9230101K08Rik; alpha 3 type IV collagen binding protein; AU016711; Ceramide transfer protein; ceramide transport protein; ceramide transporter; Cert; CERTL; Col4a3bp; collagen type IV alpha 3 binding protein; collagen type IV alpha-3-binding protein; collagen, type IV, alpha 3 (Goodpasture antigen) binding protein; goodpasture antigen-binding protein; GPBP; hCERT; lipid-transfer protein CERTL; MRD34; procollagen, type IV, alpha 3 (Goodpasture antigen) binding protein; STARD11; StAR-related lipid transfer (START) domain containing 11; stAR-related lipid transfer protein 11; START domain-containing protein 11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10087 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.71 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9Y5P4 | |
| CERT1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human COL4A3BP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction