missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERK Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25731-1-AP-150UL
This item is not returnable.
View return policy
Description
CERK has 2 isoforms with the molecular weight of 60 and 38 kDa.
CERK converts ceramide to ceramide 1-phosphate (C1P), a sphingolipid metabolite. Both CERK and C1P have been implicated in various cellular processes, including proliferation, apoptosis, phagocytosis, and inflammation (Kim et al., 2006 ).Specifications
| CERK | |
| Polyclonal | |
| Unconjugated | |
| CERK | |
| Acylsphingosine kinase, ceramide kinase, CERK, dA59H18.2, dA59H18.3, hCERK, KIAA1646, Lipid kinase 4, LK4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 223753, 64781 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8K4Q7, Q8TCT0 | |
| CERK | |
| CERK Fusion Protein Ag22774 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction