missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CERK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529306
This item is not returnable.
View return policy
Description
Recommended positive controls: H1299, HeLa, HepG2, Raji. Predicted reactivity: Mouse (88%), Rat (90%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CERK converts ceramide to ceramide 1-phosphate (C1P), a sphingolipid metabolite. Both CERK and C1P have been implicated in various cellular processes, including proliferation, apoptosis, phagocytosis, and inflammation (Kim et al., 2006 ).
Specifications
| CERK | |
| Polyclonal | |
| Unconjugated | |
| CERK | |
| acylsphingosine kinase; AI848610; Ceramide kinase; Cerk; CERK antibody; D330016D08Rik; dA59H18.2; dA59H18.3; hCERK; KIAA1646; Lipid kinase 4; lipid kinase LK4; LK4; mCERK; mKIAA1646 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64781 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.38 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q8TCT0 | |
| CERK | |
| Recombinant protein encompassing a sequence within the center region of human CERK. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction