missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CERK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109517
This item is not returnable.
View return policy
Description
Immunogen sequence: CRRSPRGLSP AAHLGDGSSD LILIRKCSRF NFLRFLIRHT NQQDQFDFTF VEVYRVKKFQ FTSKHMEDED SDLKEGGKKR FGHICSSHPS CCCTVSNSSW NCDGEVLHSP AIEVRVHCQL VRLFARGIEE NPKPDSHS.
CERK converts ceramide to ceramide 1-phosphate (C1P), a sphingolipid metabolite. Both CERK and C1P have been implicated in various cellular processes, including proliferation, apoptosis, phagocytosis, and inflammation (Kim et al., 2006 ).
Specifications
| CERK | |
| Polyclonal | |
| Unconjugated | |
| CERK | |
| acylsphingosine kinase; AI848610; Ceramide kinase; Cerk; CERK antibody; D330016D08Rik; dA59H18.2; dA59H18.3; hCERK; KIAA1646; Lipid kinase 4; lipid kinase LK4; LK4; mCERK; mKIAA1646 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 223753, 300129, 64781 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.89 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8K4Q7, Q8TCT0 | |
| CERK | |
| A synthetic peptide corresponding to a sequence within amino acids 400 to the C-terminus of human CERK (NP_073603.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction