missing translation for 'onlineSavingsMsg'
Learn More

Cerium zirconium oxide, 99.5% (metals basis), Thermo Scientific Chemicals

£135.00 - £587.00

Chemical Identifiers

CAS 53169-24-7
MDL Number MFCD00168089
Synonym Cerium zirconate
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11303618
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039216.22
100 g
£135.00
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11393608
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039216.36
500 g
£587.00
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used as wear resistant grinding and dispersion media. Used in fuel cell materials (SOFC), body armors. Also finds applications in catalytic converter, honey comb, precision ceramic parts, abrasive wheels, oxygen sensors, cutting tools. Used as ceramic steel. Other uses include screen printing, dip and spin coating and APS coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

53169-24-7
MFCD00168089
Cerium zirconate

Specifications

53169-24-7
MFCD00168089
Soluble in acids. Insoluble in water.
99.5%
Cerium zirconium oxide
Powder
Cerium zirconate
295.32
(metals basis)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 258-407-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only