missing translation for 'onlineSavingsMsg'
Learn More

Cerium tungsten oxide, 99.9% (REO), Thermo Scientific™

Product Code. 11348737
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348737 25 g 25g
11358737 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348737 Supplier Thermo Scientific Alfa Aesar Supplier No. 035840.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-74-5
Molecular Formula Ce2O9W2
Molecular Weight (g/mol) 791.91
MDL Number MFCD00049725
InChI Key PMMOMAJHUNGVIB-UHFFFAOYSA-N
Synonym cerium tungsten oxide, cerium iii tungstate, dicerium 3+ tritungstate, cerium 3+ dioxido dioxo tungsten 2/3, 2ce.3wo4, cerium tungstate ce2 wo4 3, cerous tungstate, ce2 wo4 3, dicerium 3+ ion tris dioxotungstenbis olate
PubChem CID 16213782
SMILES [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Ce+3].[Ce+3].[W].[W]

Specifications

Melting Point 1089°C
Physical Form Powder
Quantity 25 g
IUPAC Name dicerium(3+) ditungsten nonaoxidandiide
Formula Weight 1023.78
Percent Purity 99.9%
Odor Odorless
Assay (REO)
Chemical Name or Material Cerium tungsten oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.