missing translation for 'onlineSavingsMsg'
Learn More

Cerium Standard: Ce @ 1000 μg/g in Hydrocarbon Oil, VHG™

Product Code. 17962436
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962436 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17962436 Supplier VHG Labs Supplier No. VHGOCE1000A50G
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Cerium Standard: Ce @ 1000 μg/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cerium/Aluminium/Antimony/Arsenic/Barium/Beryllium/Bismuth/Boron/Cadmium/Calcium/Chromium/Cobalt/Copper/Iron/Lanthanum/Lead/Lithium/Magnesium/Manganese/Mercury/Molybdenum/Nickel/Phosphorus/Potassium/Scandium/Selenium/Silicon/Silver/Sodium/Strontium/Thallium/Tin/Titanium/Vanadium/Yttrium/Zinc/Zirconium
CAS 7440-45-1, 7429-90-5, 7440-36-0, 7440-38-2, 7440-39-3, 7440-41-7
Methods ICP/XRF/RDE
Analyte or Component Names Cerium/Aluminium/Antimony/Arsenic/Barium/Beryllium/Bismuth/Boron/Cadmium/Calcium/Chromium/Cobalt/Copper/Iron/Lanthanum/Lead/Lithium/Magnesium/Manganese/Mercury/Molybdenum/Nickel/Phosphorus/Potassium/Scandium/Selenium/Silicon/Silver/Sodium/Strontium/Thallium/Tin/Titanium/Vanadium/Yttrium/Zinc/Zirconium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix 75 cSt Mineral Oil
Quantity 50 g
Concentration or Composition (by Analyte or Components) Cerium 1000 μg/g/Aluminium (<1) μg/g/Antimony (<1) μg/g/Arsenic (<1) μg/g/Barium (<1) μg/g/Beryllium (<1) μg/g/Bismuth (<1) μg/g/Boron (<1) μg/g/Cadmium (<1) μg/g/Calcium (<1) μg/g/Chromium (<1) μg/g/Cobalt (<1) μg/g/Copper (<1) μg/g/Iron (<1) μg/g/Lanthanum (<1) μg/g/Lead (<1) μg/g/Lithium (<1) μg/g/Magnesium (<1) μg/g/Manganese (<1) μg/g/Mercury (<1) μg/g/Molybdenum (<1) μg/g/Nickel (<1) μg/g/Phosphorus (<1) μg/g/Potassium (<1) μg/g/Scandium (<1) μg/g/Selenium (<1) μg/g/Silicon (<1) μg/g/Silver (<1) μg/g/Sodium (<1) μg/g/Strontium (<1) μg/g/Thallium (<1) μg/g/Tin (<1) μg/g/Titanium (<1) μg/g/Vanadium (<1) μg/g/Yttrium (<1) μg/g/Zinc (<1) μg/g/Zirconium (<1) μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.