missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium Solution for ICP, 1g/L, HNO3 Matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£47.15 - £103.00
Specifications
Concentration | 1g/L |
---|---|
Solution Contains | HNO3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12903384
|
Fisher Chemical
S 011 100 |
100 mL |
£47.15
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12913384
|
Fisher Chemical
S 011 250 |
250 mL |
£79.90
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12923384
|
Fisher Chemical
S 011 500 |
500 mL |
£103.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
1g/L | |
HNO3 |