missing translation for 'onlineSavingsMsg'
Learn More

Cerium molybdenum oxide, 99%

Product Code. 11379357
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379357 25 g 25g
11359357 5 g 5g
11369357 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11379357 Supplier Thermo Scientific Alfa Aesar Supplier No. 040210.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13454-70-1 | Ce2Mo3O12 | 760.07 g/mol

Cerium molybdenum oxide is used in the aerospace and coating industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-70-1
Molecular Formula Ce2Mo3O12
Molecular Weight (g/mol) 760.07
MDL Number MFCD00798520
InChI Key BPOYBXJKAISYKA-UHFFFAOYSA-N
Synonym Cerium molybdate

Specifications

Melting Point 973°C
Color Brown
Physical Form Powder
Quantity 25 g
Assay Percent Range 99%
Solubility Information Insoluble in water.
Formula Weight 760.05
Chemical Name or Material Cerium molybdenum oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.