missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) trifluoromethanesulfonate, 98%

Product Code. 11314486
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314486 5 g 5g
11324486 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11314486 Supplier Thermo Scientific Alfa Aesar Supplier No. L20056.06
  • ONLINE EXCLUSIVE PRICE
    £44.50
    £40.65 / 5g
    Save £3.85
    8% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 107792-63-2 | C4CeF12O12S4 | 736.37 g/mol

Cerium(IV) trifluoromethanesulfonate, is used as a strong oxidizing agent, in the oxidation of benzyl alcohols and alkylbenzenes to aryl aldehydes and aryl ketones respectively. It can also be used as a catalyst for high-yield deprotection of tert-butyldimethylsilyl ethers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerium(IV) trifluoromethanesulfonate, is used as a strong oxidizing agent, in the oxidation of benzyl alcohols and alkylbenzenes to aryl aldehydes and aryl ketones respectively. It can also be used as a catalyst for high-yield deprotection of tert-butyldimethylsilyl ethers

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a cool, dry place in a tightly closed containers. Stable under recommended storage conditions. Incompatible with oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107792-63-2
Molecular Formula C4CeF12O12S4
Molecular Weight (g/mol) 736.37
MDL Number MFCD03844746
InChI Key BYAJTISDHIASIP-UHFFFAOYSA-J
Synonym cerium iv trifluoromethanesulfonate, cerium iv triflate, cerium trifluoromethanesulfonate, acmc-20alhh, ceric triflate, cerium iv trifluoromethanesulphonate, cerium iv trifluoromethanesulfonate, ?ce cf3so3 4, trifluoromethanesulfonic acid, cerium iv salt, methanesulfonic acid,1,1,1-trifluoro-,cerium 4+ salt 4:1
PubChem CID 9875257
IUPAC Name cerium(4+);trifluoromethanesulfonate
SMILES [Ce+4].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 736.39
Percent Purity 98%
Chemical Name or Material Cerium(IV) trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.