missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) sulfate tetrahydrate, 98+%

Product Code. 10236590 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10236590 500 g Plastic bottle 500g
10276540 100 g Plastic bottle 100g
10743894 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10236590 Supplier Thermo Scientific Acros Supplier No. 199135000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10294-42-5 | CeO8S2·4H2O | 404.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-42-5
Molecular Formula CeO8S2·4H2O
Molecular Weight (g/mol) 404.29
MDL Number MFCD00149427
InChI Key MEXSQFDSPVYJOM-UHFFFAOYSA-J
Synonym cerium iv sulfate tetrahydrate, ceric sulfate tetrahydrate, unii-6sax34b8je, 6sax34b8je, acmc-20ecws, ceric sulphate tetrahydrate, ksc493s0d, aronis24302, sulfuric acid, cerium 4+ salt 2:1 , tetrahydrate, cerium iv sulfate hydrate
PubChem CID 22723557
IUPAC Name cerium(4+);disulfate;tetrahydrate
SMILES O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+4]

Specifications

Color Yellow
Physical Form Powder
Quantity 500 g
Assay Percent Range 98% min. (Iodometry)
Linear Formula Ce(SO4)2·4H2O
Fieser 02,65; 15,73
Merck Index 14, 1990
Solubility Information Solubility in water: 38g/L (50°C). Other solubilities: soluble in dilute sulfuric acid,slowly soluble in cold,more readily soluble in,hot mineral acids
Formula Weight 404.29
Percent Purity ≥98%
Packaging Plastic bottle
Density 1.06
Chemical Name or Material Cerium(IV) sulfate tetrahydrate, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Cerium(IV) sulfate tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Supplemental information
  • MIXTURE LIST-Contain: Sulfuric acid, cerium(4+) salt (2:1), Sulfuric acid, cerium(4+) salt (2:1), tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.