missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) sulfate hydrate, REacton™, 99% (REO)

Product Code. 11331079
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331079 500 g 500g
11341079 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331079 Supplier Thermo Scientific Alfa Aesar Supplier No. 011323.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95838-16-7 | CeO8S2 | 332.23 g/mol

Cerium(IV) sulfate hydrate acts as oxidizing agent which is used for the generation of o-benzoquinone from catechol. It promotes the formation of radicals from dialkyl malonates, which undergo substitution reactions with furans and thiophenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95838-16-7
Molecular Formula CeO8S2
Molecular Weight (g/mol) 332.23
MDL Number MFCD00148852
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
Synonym sulfuricacid, cerium salt, hydrate 9ci, acmc-20mqih, cerium iv sulfate monohydrate, cerium iv sulfate hydrate, reacton, cerium 4+ sulfate-water 1/2/1
PubChem CID 20223355
IUPAC Name λ⁴-cerium(4+) hydrate disulfate
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 195°C (decomposition)
Physical Form Crystalline
Quantity 500 g
UN Number UN3085
Solubility Information Slightly soluble in water. Soluble in dil.sulfuric acid.
Formula Weight 332.24 (Anhydrous)
Percent Purity 99%
Grade Reacton
Sensitivity Moisture sensitive
Density 3.01 g/mL
Assay (REO)
Chemical Name or Material Cerium(IV) sulfate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.