missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) sulfate hydrate, 99%, ACROS Organics™

Product Code. 10194730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10194730 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10194730 Supplier Thermo Scientific Acros Supplier No. 279821000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 123333-60-8
Color Yellow
Physical Form Powder
Quantity 100 g
Assay Percent Range 99%
Molecular Formula CeH2O9S2
Linear Formula Ce(SO4)2·xH2O
Fieser 02,65; 15,73
Merck Index 14, 1990
Synonym sulfuricacid, cerium salt, hydrate 9ci, acmc-20mqih, cerium iv sulfate monohydrate, cerium iv sulfate hydrate, reacton, cerium 4+ sulfate-water 1/2/1
InChI Key KNANFSXYDCURQS-UHFFFAOYSA-J
SMILES O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+4]
IUPAC Name cerium(4+);disulfate;hydrate
Molecular Weight (g/mol) 350.243
PubChem CID 20223355
Formula Weight 332.23
Percent Purity ≥98.5% (Iodometry)
Packaging Glass bottle
Appearance (100mg/10mL 6N H2SO4) clear yellow
Chemical Name or Material Cerium(IV) sulfate hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.