missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) sulfate hydrate, 98%

Artikelnummer. 11449412
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
250 g
1000 g
missing translation for 'unitSize'
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
11449412 250 g 250g
11439412 50 g 50g
11429412 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11449412 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A10142.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 95838-16-7 | CeO8S2 | 332.23 g/mol

Cerium(IV) sulfate hydrate is used as a general application for chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerium(IV) sulfate hydrate is used as a general application for chemical intermediate.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 95838-16-7
Molecular Formula CeO8S2
Molecular Weight (g/mol) 332.23
MDL Number MFCD00148852
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
Synonym sulfuricacid, cerium salt, hydrate 9ci, acmc-20mqih, cerium iv sulfate monohydrate, cerium iv sulfate hydrate, reacton, cerium 4+ sulfate-water 1/2/1
PubChem CID 20223355
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Spezifikation

Melting Point 195°C (decomposition)
Quantity 250 g
UN Number UN3085
Merck Index 14,1990
Solubility Information Soluble in water.
IUPAC Name λ⁴-cerium(4+) disulfate
Formula Weight 332.24 (Anhydrous)
Percent Purity 98%
Density 3.01
Chemical Name or Material Cerium (IV) sulfate hydrate
compliance-icons compliance-icons
Produktidentifikator
  • Cerium(IV) sulfate hydrate
Signalwort
  • Danger
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 1 C
Gefahrenhinweis
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Cerium(IV) sulfate hydrate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.