missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) sulfate, 0.1N Standardized Solution, Thermo Scientific Chemicals

Product Code. 11353797
Click to view available options
Quantity:
1 L
4 L

Cerium(IV) sulfate is a redox indicator and is used in analytical chemistry for redox titrations. In ionized form, ceric ion acts as a strong oxidizer under acidic conditions. It is also used to prepare spherical-like nanoparticles CeO2, nanoparticles for NO reductions' reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13590-82-4
Molecular Formula CeO8S2
Molecular Weight (g/mol) 332.23
MDL Number MFCD00148852
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
PubChem CID 159684
IUPAC Name cerium(4+);disulfate
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Quantity 1 L
Color Yellow
Formula Weight 332.24
Odor Odorless
Density 1.05
Solubility Miscible with water and dilute sulfuric acid
Physical Form Liquid
Chemical Name or Material Cerium(IV) sulfate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'