missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) oxide, hydrated

Product Code. 11387997
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387997 250 g 250g
11397997 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387997 Supplier Thermo Scientific Alfa Aesar Supplier No. 021120.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12014-56-1 | CeH8O4 | 212.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12014-56-1
Molecular Formula CeH8O4
Molecular Weight (g/mol) 212.18
MDL Number MFCD00010927
InChI Key DPUCLPLBKVSJIB-UHFFFAOYSA-N
SMILES O.O.O.O.[Ce]

Specifications

Color Brown to Yellow
Physical Form Lump/Powder
Quantity 250 g
Formula Weight 172.12 (Anhydrous)
Odor Odorless
Chemical Name or Material Cerium(IV) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.