missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) isopropoxide, Ce 37-45%

Product Code. 11470790
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470790 1 g 1g
11480790 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470790 Supplier Thermo Scientific Alfa Aesar Supplier No. 044468.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63007-83-0 | C12H28CeO4 | 376.47 g/mol

Cerium(IV) isopropoxide, Ce 37-45% is used to catalyzes diastereoselective pinacol couplings. It forms diketonates ,which hydrolyze to CeO2 particles. CeO2 particles have the potential for ion conduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63007-83-0
Molecular Formula C12H28CeO4
Molecular Weight (g/mol) 376.47
MDL Number MFCD00061483
InChI Key BCBBBOXRROHVFG-UHFFFAOYSA-N
Synonym cerium iv isopropoxide, acmc-20alht, tetraisopropoxycerium iv, cerium 4+ tetrapropan-2-olate
PubChem CID 14473355
IUPAC Name cerium(4+);propan-2-olate
SMILES [Ce+4].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-]

Specifications

Melting Point 140°C (decomposition)
Color Orange-Red to Yellow
Linear Formula Ce(OC3H7)4
Quantity 1 g
UN Number UN2925
Sensitivity Air and moisture sensitive
Solubility Information Soluble in pyridine
Formula Weight 376.48
Concentration or Composition (by Analyte or Components) Ce 37 to 45%
Physical Form Powder
Chemical Name or Material Cerium (IV) isopropoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.