missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) fluoride hydrate, Thermo Scientific Chemicals

Product Code. p-7050544
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11378887

Brand: Thermo Scientific Alfa Aesar 038580.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cerium(IV) fluoride hydrates a water insoluble Cerium source for use in oxygen-sensitive applications, such as metal production. Commercial applications for cerium include metallurgy, glass and glass polishing, ceramics, catalysts, and in phosphors. In steel manufacturing it is used to remove free oxygen and sulfur by forming stable oxysulfides and by tying up undesirable trace elements, such as lead and antimony.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60627-09-0
Molecular Formula CeF4
Molecular Weight (g/mol) 216.11
MDL Number MFCD00049422
InChI Key SWURHZJFFJEBEE-UHFFFAOYSA-J
Synonym cerium iv fluoride hydrate, acmc-20alhu, cerium 4+ fluoride-water 1/4/1
PubChem CID 71317457
IUPAC Name cerium(4+);tetrafluoride;hydrate
SMILES F[Ce](F)(F)F

Specifications

Physical Form Powder
Quantity 10 g
UN Number UN3288
Solubility Information Insoluble in water; Slowly hydrolyzed by cold water
Formula Weight 216.11 (Anhydrous)
Chemical Name or Material Cerium(IV) fluoride hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.