missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) and Dilute Nitric acid, Etchant Solution, Ce(IV) concentration 0.25N, Thermo Scientific Chemicals

Product Code. 11339767
Change view
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339767 1 L 1 L
11349767 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339767 Supplier Thermo Scientific Alfa Aesar Supplier No. 042110.K2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is widely applied as etchant solution in industrial purpose to chemically remove layers from the surface of a wafer during manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD01866052

Specifications

Physical Form Liquid
Quantity 1 L
UN Number UN3098
Solubility Information Soluble in water
Concentration or Composition (by Analyte or Components) Ce(IV) concentration 0.25N
Chemical Name or Material Cerium(IV) and Dilute Nitric acid, Etchant Solution

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.