missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min

Product Code. 11339337
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339337 5 kg 5kg
11329337 250 g 250g
11349337 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339337 Supplier Thermo Scientific Alfa Aesar Supplier No. 040194.A7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10378-47-9 | (NH4)4Ce(SO4)4·2H2O

Ammonium cerium(IV) sulfate dihydrate is an oxidizing agent in organic synthesis and it is used as an analytical reagent, in thin-layer chromatography for the detection of anti-depressant drugs in human blood and urine samples. It is used to wet etch the Co catalyst and synthesis of carbon Nanotubes. It is used in medical and pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10378-47-9
Molecular Formula (NH4)4Ce(SO4)4·2H2O
MDL Number MFCD00149417
Synonym Ammonium cerium sulfate

Specifications

Melting Point 130°C
Color Yellow
Physical Form Crystalline
Quantity 5 kg
Assay Percent Range 94% min.
Solubility Information Soluble in water and insoluble in ethanol.
Formula Weight 632.56 (596.53 Anhydrous)
Grade ACS
Appearance Yellow crystalline
Chemical Name or Material Cerium(IV) ammonium sulfate dihydrate, For analysis ACS

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.