Learn More
Cerium(IV) ammonium nitrate, ACS, 98.5% min
Description
Analytical chemistry, as an oxidant for organic compounds, as a polymerization catalyst for olefins, and as a scavenger in the manufacture of azides. Cerium(IV) ammonium nitrate is widely utilized as an oxidant for many functional groups like alcohols, phenols and ethers as well as C-H bonds. It acts as a deprotection reagent for alcohol in organic synthesis. It is useful in the synthesis of heterocycles like quinoxaline derivatives which is used in dyes, organic semiconductors and DNA cleaving agents. It is an active component of chrome etchant, which is used in the production of photomasks and liquid crystal displays.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Granules |
| Quantity | 500 g |
| Assay Percent Range | 98.5% |
| Solubility Information | Soluble in water and alcohol. |
| Formula Weight | 548.23 |
| Grade | ACS |
| Chemical Name or Material | Cerium(IV) ammonium nitrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.