Learn More
Cerium(III) sulfate octahydrate, REacton™, 99.999% (metals basis), Thermo Scientific Chemicals
£129.00 - £491.00
Chemical Identifiers
CAS | 10450-59-6 |
---|---|
Molecular Formula | Ce2(SO4)3·8H2O |
MDL Number | MFCD00149641 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11458510
|
Thermo Scientific Alfa Aesar
044357.18 |
50 g |
£129.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468510
|
Thermo Scientific Alfa Aesar
044357.30 |
250 g |
£491.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cerium(III) sulfate octahydrate used in waterproofing, mildew-proofing, and in dyeing and printing textiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10450-59-6 | |
Ce2(SO4)3·8H2O | |
MFCD00149641 |
Specifications
10450-59-6 | |
Ce2(SO4)3·8H2O | |
14,2002 | |
712.55 (568.42 Anhydrous) | |
Reacton™ | |
(metals basis) |
Crystalline | |
MFCD00149641 | |
Soluble in water. | |
99.999% | |
2.886 g/mL | |
Cerium (III) sulfate octahydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only