missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) sulfate, 99.9%, (REO=Rare Earth Oxides), Thermo Scientific Chemicals

Catalog Number 10046182
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13454-94-9
Ce2O12S3
568.43
MFCD00010931
OZECDDHOAMNMQI-UHFFFAOYSA-H
cerium sulfate, cerous sulfate, cerium iii sulfate, cerium sulphate, dicerium trisulfate, cerium sesquisulfate, unii-0vf70o14r1, sulfuric acid, cerium salt, dicerium trisulphate, sulphuric acid, cerium salt
159674
cerium(3+);trisulfate
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+3].[Ce+3]
This item is not returnable. View return policy
13454-94-9
Ce2O12S3
568.43
MFCD00010931
OZECDDHOAMNMQI-UHFFFAOYSA-H
cerium sulfate, cerous sulfate, cerium iii sulfate, cerium sulphate, dicerium trisulfate, cerium sesquisulfate, unii-0vf70o14r1, sulfuric acid, cerium salt, dicerium trisulphate, sulphuric acid, cerium salt
159674
cerium(3+);trisulfate
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+3].[Ce+3]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY
Color Colorless
Physical Form Liquid
Quantity 50 g
Assay Percent Range 46.0 to 50% (Ce) (Complexometry)
Linear Formula Ce2(SO4)3
Merck Index 15, 2001
Solubility Information Solubility in water: slightly soluble
Formula Weight 568.43
Percent Purity 99.90%
Packaging Glass bottle
Chemical Name or Material Cerium(III) sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'