missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) phosphate, 99% min

Product Code. 11383608
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383608 25 g 25g
11373608 100 g 100g
11363608 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11383608 Supplier Thermo Scientific Alfa Aesar Supplier No. 039215.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13454-71-2 | CeO4P | 235.09 g/mol

Intermediate product in cerium-based technique for staining lysosomal enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-71-2
Molecular Formula CeO4P
Molecular Weight (g/mol) 235.09
MDL Number MFCD00049424
InChI Key TYAVIWGEVOBWDZ-UHFFFAOYSA-K
Synonym cerium phosphate, cerium 3+ phosphate, unii-xr1z4biw2j, cerium iii phosphate, xr1z4biw2j, phosphoric acid,cerium 3+ salt 1:1, cerium monophosphate, cerium 3 phosphate, cerium orthophosphate cepo4, acmc-20alhk
PubChem CID 159673
IUPAC Name cerium(3+);phosphate
SMILES [Ce+3].[O-]P([O-])([O-])=O

Specifications

Color Yellow
Physical Form Powder
Quantity 25 g
Linear Formula CePO4
Solubility Information Insoluble in water.
Formula Weight 235.09
Percent Purity ≤99%
Odor Odorless
Density 5.22 g/mL
Chemical Name or Material Cerium(III) phosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.