missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) oxalate hydrate, REacton™, 99.989% (REO), Thermo Scientific™

Product Code. 11336678
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336678 50 g 50g
11326678 10 g 10g
11316678 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11336678 Supplier Thermo Scientific Alfa Aesar Supplier No. 013226.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15750-47-7
Molecular Formula C6Ce2O12
Molecular Weight (g/mol) 544.29
MDL Number MFCD00013082
InChI Key ZMZNLKYXLARXFY-UHFFFAOYSA-H
Synonym cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate
PubChem CID 91998046
SMILES [Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Specifications

Melting Point (decomposition)
Physical Form Crystalline
Quantity 50 g
UN Number UN3288
Merck Index 14,2001
IUPAC Name cerium;oxalic acid;hydrate
Formula Weight 544.29 (Anhydrous)
Percent Purity 99.989%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Cerium(III) oxalate hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.