missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) nitrate hexahydrate, REacton™, 99.99% (REO)

Product Code. 11311099
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311099 50 g 50g
11321099 5 g 5g
11301099 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11311099 Supplier Thermo Scientific Alfa Aesar Supplier No. 011330.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10294-41-4 | CeH12N3O15 | 434.22 g/mol

Cerium(III) nitrate hexahydrate is used in the separation of cerium from other rare earths. It plays an important role as a catalyst for hydrolysis of phosphoric acid esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-41-4
Molecular Formula CeH12N3O15
Molecular Weight (g/mol) 434.22
MDL Number MFCD00149631
InChI Key QQZMWMKOWKGPQY-UHFFFAOYSA-N
Synonym cerium iii nitrate hexahydrate, cerium nitrate hexahydrate, cerous nitrate hexahydrate, unii-fl1r38jebb, fl1r38jebb, nitric acid cerium salt, cerium iii nitratehexahydrate, ce.3no3.6h2o, ksc494m5f
PubChem CID 16211466
IUPAC Name cerium(3+) hexahydrate trinitrate
SMILES O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 50 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in water,alcohol and acetone.
Formula Weight 434.23 (326.13 Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Cerium(III) nitrate hexahydrate, hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.