missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific™

Product Code. 11386918
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386918 250 g 250g
11396918 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386918 Supplier Thermo Scientific Alfa Aesar Supplier No. 013662.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-88-5
Molecular Formula CeF3
Molecular Weight (g/mol) 197.111
MDL Number MFCD00010932
InChI Key QCCDYNYSHILRDG-UHFFFAOYSA-K
Synonym cerium fluoride, cerous fluoride, cerium 3+ trifluoride, cerium fluorure french, unii-1gct2g09an, cerium +3 cation trifluoride, 1gct2g09an, cerium fluorure, acmc-20alhs
PubChem CID 24457
IUPAC Name cerium(3+);trifluoride
SMILES [F-].[F-].[F-].[Ce+3]

Specifications

Boiling Point ∼2300°C
Melting Point 1460°C
Physical Form Lump
Quantity 250 g
Merck Index 14,1998
Solubility Information Soluble in strong mineral acids.
Formula Weight 197.12
Percent Purity 99.9%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Density 6.16 g/mL
Assay (REO)
Chemical Name or Material Cerium(III) fluoride, Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.