missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) fluoride, 99.9% (REO)

Product Code. 11357997
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357997 500 g 500g
11377997 25 g 25g
11367997 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11357997 Supplier Thermo Scientific Alfa Aesar Supplier No. 021118.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7758-88-5 | CeF3 | 197.111 g/mol

Cerium(III) fluoride is used to make Lewis acid. It is considered to be the most effective glass polishing agent for high precision optical polishing. Cerium fluoride used as a core additive to improve the properties of the eventual weld. The fluoride provides stability to the arc and also modifies the behavior of the molten metal flowing into the weld.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-88-5
Molecular Formula CeF3
Molecular Weight (g/mol) 197.111
MDL Number MFCD00010932
InChI Key QCCDYNYSHILRDG-UHFFFAOYSA-K
Synonym cerium fluoride, cerous fluoride, cerium 3+ trifluoride, cerium fluorure french, unii-1gct2g09an, cerium +3 cation trifluoride, 1gct2g09an, cerium fluorure, acmc-20alhs
PubChem CID 24457
IUPAC Name cerium(3+);trifluoride
SMILES [F-].[F-].[F-].[Ce+3]

Specifications

Boiling Point ∼2300°C
Melting Point 1460°C
Physical Form Powder
Quantity 500 g
Merck Index 14,1998
Solubility Information Practically insoluble in, but slowly hydrolyzed by water
Formula Weight 197.12
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 6.16 g/mL
Assay (REO)
Chemical Name or Material Cerium(III) fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.