Learn More
Cerium(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
£84.40 - £621.00
Chemical Identifiers
CAS | 7790-86-5 |
---|---|
Molecular Formula | CeCl3 |
Molecular Weight (g/mol) | 246.466 |
MDL Number | MFCD00010929 |
InChI Key | VYLVYHXQOHJDJL-UHFFFAOYSA-K |
Synonym | cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride |
PubChem CID | 24636 |
ChEBI | CHEBI:35458 |
IUPAC Name | cerium(3+);trichloride |
SMILES | [Cl-].[Cl-].[Cl-].[Ce+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11343867
|
Thermo Scientific Alfa Aesar
035688.06 |
5 g |
£84.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11333867
|
Thermo Scientific Alfa Aesar
035688.14 |
25 g |
£312.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11405050
|
Thermo Scientific Alfa Aesar
035688.22 |
100 g |
£621.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cerium(III) chloride is used in incandescent gas mantles, spectrography and polymerization of olefins. It acts as a Lewis acid and used in Friedel-Crafts alkylation and acylation reactions in organic synthesis. It is used as a precursor for the preparation of other cerium salts, such as cerium(III) trifluoromethanesulfonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7790-86-5 | |
246.466 | |
VYLVYHXQOHJDJL-UHFFFAOYSA-K | |
24636 | |
cerium(3+);trichloride |
CeCl3 | |
MFCD00010929 | |
cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride | |
CHEBI:35458 | |
[Cl-].[Cl-].[Cl-].[Ce+3] |
Specifications
1727°C | |
7790-86-5 | |
Crystalline | |
CeCl3 | |
cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride | |
VYLVYHXQOHJDJL-UHFFFAOYSA-K | |
cerium(3+);trichloride | |
24636 | |
246.48 | |
Hygroscopic | |
3.92 g/cm3 at 20°C |
848°C | |
White | |
99.9% | |
MFCD00010929 | |
Soluble in water,ethanol,acids and acetone. | |
[Cl-].[Cl-].[Cl-].[Ce+3] | |
246.466 | |
CHEBI:35458 | |
Odorless | |
White crystalline beads | |
Cerium(III) chloride |
Safety and Handling
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 232-227-8
missing translation for 'rtecsNumber' : FK5075000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Hygroscopic
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only