missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) chloride hydrate, REacton™, 99.9% (REO)

Product Code. 11391079
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391079 250 g 250g
11381079 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391079 Supplier Thermo Scientific Alfa Aesar Supplier No. 011325.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19423-76-8 | CeCl3 | 246.47 g/mol

Cerium(III) chloride hydrate is used as an important raw material the preparation of other cerium salts such as cerium(III) trifluoromethanesulfonate. It is used in the conversion of esters to allylsilanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19423-76-8
Molecular Formula CeCl3
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149633
InChI Key VYLVYHXQOHJDJL-UHFFFAOYSA-K
Synonym Cerous chloride
IUPAC Name cerium(3+) hydrate trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ce+3]

Specifications

Color White
Physical Form Crystalline Aggregates
Quantity 250 g
Assay Percent Range 99.9%
Solubility Information Soluble in water and strong mineral acids.
Formula Weight 246.48 (Anhydrous)
Sensitivity Air sensitive; Hygroscopic
Chemical Name or Material Cerium(III) chloride hydrate, Rare Earth Oxides Content

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.