missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific™

Product Code. 15437465
Click to view available options
Quantity:
1g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14457-87-5
MDL Number MFCD00016004

Specifications

Quantity 5g
Merck Index 14,1995
Solubility Information Soluble in water.
Formula Weight 379.83
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 5.18 g/mL
Assay (metals basis)
Chemical Name or Material Cerium(III) bromide, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'