missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) bromide hydrate, 99.998% (metals basis)

Product Code. 11410730
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410730 10 g 10g
11420730 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410730 Supplier Thermo Scientific Alfa Aesar Supplier No. 044432.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14457-87-5 | Br3Ce | 379.83 g/mol

Cerium(III) bromide is used in the following ways, CeBr3-doped lanthanum bromide single crystals are known to exhibit superior scintillation properties for applications in the security, medical imaging, and geophysics detectors.Undoped single crystals of CeBr3 have shown promise as a γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14457-87-5
Molecular Formula Br3Ce
Molecular Weight (g/mol) 379.83
MDL Number MFCD00016004
InChI Key MOOUSOJAOQPDEH-UHFFFAOYSA-K
IUPAC Name cerium(3+) tribromide
SMILES [Br-].[Br-].[Br-].[Ce+3]

Specifications

Color Orange
Physical Form Crystalline
Quantity 10 g
Merck Index 14,1995
Solubility Information Soluble in water.
Formula Weight 379.83 (Anhydrous)
Percent Purity 99.998%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Cerium (III) bromide hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.